missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(dimethylamino)benzophenone 98.0+%, TCI America™

Catalog No. B0481500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0481500G 500 g
B048125G 25 g
B0481100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0481500G Supplier TCI America Supplier No. B0481500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90-94-8
Molecular Formula C17H20N2O
Molecular Weight (g/mol) 268.36
MDL Number MFCD00008312
InChI Key VVBLNCFGVYUYGU-UHFFFAOYSA-N
Synonym michler's ketone, 4,4'-bis dimethylamino benzophenone, michler ketone, bis 4-dimethylamino phenyl methanone, p,p'-michler's ketone, michlers ketone, methanone, bis 4-dimethylamino phenyl, tetramethyldiaminobenzophenone, p,p'-bis dimethylamino benzophenone
PubChem CID 7031
ChEBI CHEBI:82347
IUPAC Name 4-[4-(dimethylamino)benzoyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 174°C
Boiling Point 360°C
Quantity 500 g
Formula Weight 268.36
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(dimethylamino)benzophenone
EINECSNumber (4)-0892
RTECSNumber DJ0250000
TSCA Yes