missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis[4-(di-p-tolylamino)styryl]biphenyl 98.0+%, TCI America™

Catalog No. B4682200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4682200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4682200MG Supplier TCI America Supplier No. B4682200MG
Only null left

Chemical Identifiers

CAS 119586-44-6
Molecular Formula C56H48N2
Molecular Weight (g/mol) 749.014
MDL Number MFCD09751239
InChI Key OSQXTXTYKAEHQV-WXUKJITCSA-N
Synonym 4,4′C-Bis[2-[4-[N,N-di(p-tolyl)amino]phenyl]vinyl]biphenyl
PubChem CID 14366516
IUPAC Name 4-methyl-N-[4-[(E)-2-[4-[4-[(E)-2-[4-(4-methyl-N-(4-methylphenyl)anilino)phenyl]ethenyl]phenyl]phenyl]ethenyl]phenyl]-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)C5=CC=C(C=C5)C=CC6=CC=C(C=C6)N(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C

Specifications

Quantity 200 mg
Formula Weight 749.01
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis[4-(di-p-tolylamino)styryl]biphenyl