missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(2,2-diphenylvinyl)biphenyl (purified by sublimation) 99.0+%, TCI America™

Catalog No. B5139200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5139200MG 200 mg
B51391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5139200MG Supplier TCI America Supplier No. B5139200MG
Only null left

Chemical Identifiers

CAS 142289-08-5
Molecular Formula C40H30
Molecular Weight (g/mol) 510.68
MDL Number MFCD03093280
InChI Key UHXOHPVVEHBKKT-UHFFFAOYSA-N
Synonym DPVBi
PubChem CID 15475895
IUPAC Name 1-(2,2-diphenylethenyl)-4-[4-(2,2-diphenylethenyl)phenyl]benzene
SMILES C1=CC=C(C=C1)C(=CC2=CC=C(C=C2)C3=CC=C(C=C3)C=C(C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Melting Point 209°C
Color Yellow
Quantity 200 mg
Formula Weight 510.68
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(2,2-diphenylvinyl)biphenyl (purified by sublimation)