missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Aminopiperidino)pyridine Dihydrochloride 98.0+%, TCI America™

Numéro de catalogue. A16551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A16551G 1 g
A16555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A16551G Fournisseur TCI America Code fournisseur A16551G
Il en reste null

Identifiants chimiques

CAS 1169396-92-2
Molecular Formula C10H17Cl2N3
Molecular Weight (g/mol) 250.167
MDL Number MFCD06797043
InChI Key YIMBYMVSBQILFO-UHFFFAOYSA-N
Synonym 4-Amino-1-(4-pyridyl)piperidine Dihydrochloride
PubChem CID 44630068
IUPAC Name 1-pyridin-4-ylpiperidin-4-amine;dihydrochloride
SMILES C1CN(CCC1N)C2=CC=NC=C2.Cl.Cl

Spécifications

Color White-Yellow
Quantity 1 g
Formula Weight 250.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Aminopiperidino)pyridine Dihydrochloride