missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane 97.0+%, TCI America™

Catalog No. T33225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33225G 5 g
T33221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33225G Supplier TCI America Supplier No. T33225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 890042-13-4
Molecular Formula C24H23BO2
Molecular Weight (g/mol) 354.26
MDL Number MFCD26401844
InChI Key VXLBBSLCTFTKOE-UHFFFAOYSA-N
Synonym 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)triphenylene, Triphenylen-2-ylboronic Acid Pinacol Ester
PubChem CID 58769449
IUPAC Name 4,4,5,5-tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 5 g
Formula Weight 354.26
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane