missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-1,3,2-dioxaphospholane 2-Oxide 95.0+%, TCI America™

Catalog No. T26455G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26455G 5 g
T26451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26455G Supplier TCI America Supplier No. T26455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16352-18-4
Molecular Formula C6H12O3P+
Molecular Weight (g/mol) 163.133
MDL Number MFCD09836231
InChI Key MWVBQXVOGPKEII-UHFFFAOYSA-N
PubChem CID 11309699
IUPAC Name 4,4,5,5-tetramethyl-1,3,2-dioxaphospholan-2-ium 2-oxide
SMILES CC1(C(O[P+](=O)O1)(C)C)C

Specifications

Melting Point 104°C
Color White
Quantity 5 g
Formula Weight 164.14
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,5,5-Tetramethyl-1,3,2-dioxaphospholane 2-Oxide