missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Tris(benzoyloxy)trityl Bromide 97.0+%, TCI America™

Catalog No. T10715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10715G 5 g
T107125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10715G Supplier TCI America Supplier No. T10715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86610-66-4
Molecular Formula C40H27BrO6
Molecular Weight (g/mol) 683.554
MDL Number MFCD00010302
InChI Key PKXTVZCJOBWKIG-UHFFFAOYSA-N
Synonym Tris[4-(phenylcarbonyloxy)phenyl]methyl Bromide
PubChem CID 3933738
IUPAC Name [4-[bis(4-benzoyloxyphenyl)-bromomethyl]phenyl] benzoate
SMILES C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)C(C3=CC=C(C=C3)OC(=O)C4=CC=CC=C4)(C5=CC=C(C=C5)OC(=O)C6=CC=CC=C6)Br

Specifications

Melting Point 190°C
Quantity 5 g
Formula Weight 683.55
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′,4′′-Tris(benzoyloxy)trityl Bromide [Hydroxyl Protecting Agent]