missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione 98.0+%, TCI America™

Catalog No. T15835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15835G 5 g
T158325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15835G Supplier TCI America Supplier No. T15835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 893-33-4
Molecular Formula C14H9F3O2
Molecular Weight (g/mol) 266.219
MDL Number MFCD00054501
InChI Key WVVLURYIQCXPIV-UHFFFAOYSA-N
Synonym 4,4,4-trifluoro-1-naphthalen-2-yl butane-1,3-dione, 4,4,4-trifluoro-1-2-naphthyl-1,3-butanedione, 1-2-naphthoyl-3,3,3-trifluoroacetone, 4,4,4-trifluoro-1-2-naphthyl butane-1,3-dione, 3-2-naphthoyl-1,1,1-trifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-naphthalenyl, 2-naphthoyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-naphthyl, 4,4,4-trifluoro-3-oxo-2'-butyronaphthone, acmc-1bl5t
PubChem CID 70179
IUPAC Name 4,4,4-trifluoro-1-naphthalen-2-ylbutane-1,3-dione
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)CC(=O)C(F)(F)F

Specifications

Melting Point 74°C
Quantity 5 g
Formula Weight 266.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione