missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Tri-9-carbazolyltriphenylamine (purified by sublimation) 98.0+%, TCI America™

Catalog No. T2274200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2274200MG 200 mg
T22741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2274200MG Supplier TCI America Supplier No. T2274200MG

Chemical Identifiers

CAS 139092-78-7
Molecular Formula C54H36N4
Molecular Weight (g/mol) 740.91
MDL Number MFCD03093250
InChI Key AWXGSYPUMWKTBR-UHFFFAOYSA-N
Synonym tris 4-9h-carbazol-9-yl phenyl amine, tcta, tcata, 4,4',4-tris carbazol-9-yl-triphenylamine, 4,4',4-tri-9-carbazolyltriphenylamine, tris 4-carbazoyl-9-ylphenyl amine, tris 4-carbazol-9-ylphenyl amine, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-yl phenyl aniline, 4,4',4-tris carbaz, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-ylphenyl aniline
PubChem CID 9962045
IUPAC Name 4-(9H-carbazol-9-yl)-N,N-bis[4-(9H-carbazol-9-yl)phenyl]aniline
SMILES C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=C(C=C1)N(C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 266°C
Color White-Yellow
Quantity 200 mg
Formula Weight 740.91
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′,4′′-Tri-9-carbazolyltriphenylamine (purified by sublimation)