missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole 97.0+%, TCI America™

Catalog No. T27565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27565G 5 g
T27561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27565G Supplier TCI America Supplier No. T27565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 269410-08-4
Molecular Formula C9H15BN2O2
Molecular Weight (g/mol) 194.04
MDL Number MFCD03453063
InChI Key TVOJIBGZFYMWDT-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, pyrazole-4-boronic acid pinacol ester, 4-pyrazoleboronic acid pinacol ester, 1h-pyrazole-4-boronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole-4-boronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-1h-pyrazol-4-yl-1,3,2-dioxaborolane, pyrazole-4-boronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 2-4-pyrazolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 2774010
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
SMILES CC1(C)OB(OC1(C)C)C1=CNN=C1

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 5 g
Formula Weight 194.04
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole