missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine 98.0+%, TCI America™

Catalog No. T34775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34775G 5 g
T34771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34775G Supplier TCI America Supplier No. T34775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 364794-79-6
Molecular Formula C17H26BNO3
Molecular Weight (g/mol) 303.21
MDL Number MFCD04974052
InChI Key JOIXYIWXEYXHHG-UHFFFAOYSA-N
Synonym 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 4-morpholinomethyl phenylboronic acid pinacol ester, 4-4-morpholinomethyl phenylboronic acid pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 4-methylmorpholino phenylboronic acid, pinacol ester, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 4-morpholinomethyl phenylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-morpholin-4-ylmethyl phenyl-1,3,2-dioxaborolane
PubChem CID 2795502
IUPAC Name 4-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}morpholine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCOCC2)C=C1

Specifications

Melting Point 89°C
Color White
Quantity 5 g
Formula Weight 303.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine