missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Alcohol 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 200 mg |
| Formula Weight | 216.16 |
| Percent Purity | ≥97.0% (HPLC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Alcohol |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |