missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 125°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 230.15 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |