missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Methyl-1,2,4-oxadiazol-5-yl)benzaldehyde, 90%, Thermo Scientific™

Numéro de catalogue. CC50904CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC50904CB 250mg Amber Glass Bottle
CC50904DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC50904CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC50904CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 876316-27-7
Formule moléculaire C10H8N2O2
Poids moléculaire (g/mol) 188.186
Numéro MDL MFCD09025865
Clé InChI AEMXYVCSLPLJOW-UHFFFAOYSA-N
Synonyme 4-3-methyl-1,2,4-oxadiazol-5-yl benzaldehyde, 3-methyl-5-4-formylphenyl-1,2,4-oxadiazole, benzaldehyde,4-3-methyl-1,2,4-oxadiazol-5-yl
PubChem CID 18525855
Nom de l’IUPAC 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzaldehyde
SOURIRES CC1=NOC(=N1)C2=CC=C(C=C2)C=O

Spécifications

Nom ou substance chimique 4-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzaldehyde
CAS % minimum 89.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 90%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 188.19
Pourcentage de pureté 90%
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.