missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Dimethylamino)propoxy]-N-methylbenzylamine, 90%, Thermo Scientific™

Chemical Identifiers

CAS 910037-04-6
Molecular Formula C13H22N2O
Molecular Weight (g/mol) 222.332
MDL Number MFCD09064994
InChI Key LFLFEMOINFHORX-UHFFFAOYSA-N
Synonym 4-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-4-methylamino methyl phenoxy propyl amine, n,n-dimethyl-3-4-methylamino methyl phenoxy propan-1-amine, 4-3-dimethylamino propoxy phenyl methyl methyl amine, benzenemethanamine, 4-3-dimethylamino propoxy-n-methyl
PubChem CID 24229610
IUPAC Name N,N-dimethyl-3-[4-(methylaminomethyl)phenoxy]propan-1-amine
SMILES CNCC1=CC=C(C=C1)OCCCN(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC59546CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC59546DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

910037-04-6
222.332
LFLFEMOINFHORX-UHFFFAOYSA-N
24229610
CNCC1=CC=C(C=C1)OCCCN(C)C
C13H22N2O
MFCD09064994
4-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-4-methylamino methyl phenoxy propyl amine, n,n-dimethyl-3-4-methylamino methyl phenoxy propan-1-amine, 4-3-dimethylamino propoxy phenyl methyl methyl amine, benzenemethanamine, 4-3-dimethylamino propoxy-n-methyl
N,N-dimethyl-3-[4-(methylaminomethyl)phenoxy]propan-1-amine

Specifications

910037-04-6
100.0
C13H22N2O
4-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-4-methylamino methyl phenoxy propyl amine, n,n-dimethyl-3-4-methylamino methyl phenoxy propan-1-amine, 4-3-dimethylamino propoxy phenyl methyl methyl amine, benzenemethanamine, 4-3-dimethylamino propoxy-n-methyl
CNCC1=CC=C(C=C1)OCCCN(C)C
222.332
222.33
4-[3-(Dimethylamino)propoxy]-N-methylbenzylamine
89.5
90%
MFCD09064994
LFLFEMOINFHORX-UHFFFAOYSA-N
N,N-dimethyl-3-[4-(methylaminomethyl)phenoxy]propan-1-amine
24229610
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents