missing translation for 'onlineSavingsMsg'
Learn More

4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline 98.0+%, TCI America™

Catalog No. C29655G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29655G 5 g
C29651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29655G Supplier TCI America Supplier No. C29655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 231278-20-9
Molecular Formula C21H14ClFIN3O
Molecular Weight (g/mol) 505.72
MDL Number MFCD09998827
InChI Key UHFPFDMMKYQMLC-UHFFFAOYSA-N
Synonym n-3-chloro-4-3-fluorobenzyl oxy phenyl-6-iodoquinazolin-4-amine, n-3-chloro-4-3-fluorobenzyloxy phenyl-6-iodoquinazolin-4-amine, n-3-chloro-4-3-fluorobenzyloxy-phenyl-6-iodoquinazolin-4-amine, n-3-chloro-4-3-fluorophenyl methoxy phenyl-6-iodoquinazolin-4-amine, 6-iodo-4-3-chloro-4-3-fluorobenzyloxy anilino quinazoline, 4-3-chloro-4-3-fluorobenzyloxy phenylamino-6-iodoquinazoline, n-3-chloro-4-3-fluorobenzyl oxy phenyl-6-iodo-4-quinazolinamine, 4-quinazolinamine, n-3-chloro-4-3-fluorophenyl methoxy phenyl-6-iodo
PubChem CID 10174519
IUPAC Name N-{3-chloro-4-[(3-fluorophenyl)methoxy]phenyl}-6-iodoquinazolin-4-amine
SMILES FC1=CC=CC(COC2=C(Cl)C=C(NC3=C4C=C(I)C=CC4=NC=N3)C=C2)=C1

Specifications

Melting Point 225°C
Color White-Yellow
Quantity 5 g
Formula Weight 505.72
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline