missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Aminophenyl)-2-methyl-3-butyn-2-ol 98.0+%, TCI America™

Catalog No. A16075G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16075G Supplier TCI America Supplier No. A16075G
Only null left

Chemical Identifiers

CAS 69088-96-6
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD06200785
InChI Key DQPSETABKZMTEZ-UHFFFAOYSA-N
Synonym 3-(3-Hydroxy-3-methyl-1-butyn-1-yl)aniline
PubChem CID 112236
IUPAC Name 4-(3-aminophenyl)-2-methylbut-3-yn-2-ol
SMILES CC(C)(C#CC1=CC(=CC=C1)N)O

Specifications

Melting Point 120°C
Quantity 5 g
Formula Weight 175.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(3-Aminophenyl)-2-methyl-3-butyn-2-ol