missing translation for 'onlineSavingsMsg'
Learn More

4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzonitrile, 97%, Thermo Scientific™

Numéro de catalogue. CC68216DA
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC68216DA 1g Amber Glass Bottle
CC68216DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC68216DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC68216DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 56935-79-6
Formule moléculaire C12H11N3
Poids moléculaire (g/mol) 197.241
Numéro MDL MFCD09817559
Clé InChI BTXLPDNFEZIQBZ-UHFFFAOYSA-N
Synonyme 4-3,5-dimethyl-1h-pyrazol-1-yl benzonitrile, 4-3,5-dimethylpyrazol-1-yl benzonitrile, acmc-209luz, 4-3,5-dimethylpyrazol-1-yl-benzonitrile, 4-3,5-dimethylpyrazolyl benzenecarbonitrile, 1-4-cyanophenyl-3,5-dimethyl-1h-pyrazole, benzonitrile, 4-3,5-dimethyl-1h-pyrazol-1-yl
PubChem CID 15005387
Nom de l’IUPAC 4-(3,5-dimethylpyrazol-1-yl)benzonitrile
SOURIRES CC1=CC(=NN1C2=CC=C(C=C2)C#N)C

Spécifications

Nom ou substance chimique 4-(3, 5-Dimethyl-1H-pyrazol-1-yl)benzonitrile
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 197.24
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.