missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine 98.0+%, TCI America™

Catalog No. T32371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T32371G Supplier TCI America Supplier No. T32371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 852227-95-3
Molecular Formula C16H24BNO3
Molecular Weight (g/mol) 289.18
MDL Number MFCD03412097
InChI Key NCJDKFFODGZRRL-UHFFFAOYSA-N
Synonym 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-morpholinophenylboronic acid pinacol ester, 3-morpholino phenylboronic acid pinacol ester, 3-4-morpholino phenylboronic acid pinacol ester, 4-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, pubchem19450, ksc496k1f, 3-morpholinophenylboronic acidpinacolester, 3-morpholinophenylboronic acid pinacol
PubChem CID 4192663
IUPAC Name 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
SMILES CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)N1CCOCC1

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 1 g
Formula Weight 289.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine