missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Trifluoromethyl-benzoimidazol-1-yl)-butyric acid, ≥97%, Thermo Scientific™

Numéro de catalogue. MO07743DA
Modifier l'affichage
Click to view available options
Quantité:
1g
Emballage:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
MO07743DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. MO07743DA Fournisseur Thermo Scientific Chemicals Code fournisseur MO07743DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 876728-42-6
Formule moléculaire C12H11F3N2O2
Poids moléculaire (g/mol) 272.227
Numéro MDL MFCD07397470
Clé InChI HQUSGKZLJZMIHU-UHFFFAOYSA-N
Synonyme 4-2-trifluoromethyl-1h-benzimidazol-1-yl butanoic acid, 4-2-trifluoromethyl-benzoimidazol-1-yl-butyric acid, 4-2-trifluoromethyl benzimidazolyl butanoic acid, 1h-benzimidazole-1-butanoicacid, 2-trifluoromethyl, 4-2-trifluoromethyl benzimidazol-1-yl butanoic acid, 4-2-trifluoromethyl-1,3-benzodiazol-1-yl butanoic acid, 4-2-trifluoromethyl-1h-benzimidazol-1-yl-butanoic acid, 4-2-trifluoromethyl-1-benzimidazolyl butanoic acid, 4-2-trifluoromethyl-1h-benzo d imidazol-1-yl butanoic acid
PubChem CID 6494232
Nom de l’IUPAC 4-[2-(trifluoromethyl)benzimidazol-1-yl]butanoic acid
SOURIRES C1=CC=C2C(=C1)N=C(N2CCCC(=O)O)C(F)(F)F

Spécifications

Nom ou substance chimique 4-[2-(Trifluoromethyl)-1H-benzimidazol-1-yl]butanoic acid
CAS % minimum 97.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 272.22
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.