missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Thienyl)piperidine 98.0+%, TCI America™

Numéro de catalogue. T3107200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T3107200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. T3107200MG Fournisseur TCI America Code fournisseur T3107200MG
Il en reste null

Identifiants chimiques

CAS 198334-38-2
Molecular Formula C9H13NS
Molecular Weight (g/mol) 167.27
MDL Number MFCD12027113
InChI Key SLQVNTKKWCOXKW-UHFFFAOYSA-N
Synonym 2-(4-Piperidyl)thiophene
PubChem CID 19095574
IUPAC Name 4-(thiophen-2-yl)piperidine
SMILES C1CC(CCN1)C1=CC=CS1

Spécifications

Melting Point 48°C
Quantity 200 mg
Formula Weight 167.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-(2-Thienyl)piperidine