missing translation for 'onlineSavingsMsg'
Learn More

4-(2'-Pyridylazo)resorcinol, 97+%, ACS reagent

Catalog No. AC452930050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452930050 5 g
AC452930010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452930050 Supplier Thermo Scientific Chemicals Supplier No. 452930050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1141-59-9 | C11H9N3O2 | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1141-59-9
Molecular Formula C11H9N3O2
Molecular Weight (g/mol) 215.21
InChI Key VLCAILLZPUINNF-LCYFTJDESA-N
Synonym 4-2-pyridylazo resorcinol, 4-2-pyridlyazo resorcinol, 4-2-pyridylazo-2-resorcinol, resorcinol, 4-2-pyridylazo, 1-2-pyridylazo resorcinol, unii-n68t40h95t, 1,3-benzenediol, 4-2-pyridinylazo, 4-pyridin-2-yldiazenyl benzene-1,3-diol, 4-2-pyridylazo-1,3-benzenediol, 1,3-benzenediol, 4-2-2-pyridinyl diazenyl
PubChem CID 5474737
IUPAC Name (4Z)-3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES C1=CC=NC(=C1)NN=C2C=CC(=O)C=C2O

Specifications

Melting Point 188.0°C
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 5 g
Merck Index 15,7125
Formula Weight 215.21
Percent Purity 97+%
Clarity Passes test
Grade ACS Reagent
Chemical Name or Material 4-(2'-Pyridylazo)resorcinol
Hazard Category GHS Signal Word: Warning
EINECSNumber 214-528-6

RUO – Research Use Only