missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Naphthyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. N0946200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0946200MG 200 mg
N09461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0946200MG Supplier TCI America Supplier No. N0946200MG

Chemical Identifiers

CAS 918655-03-5
Molecular Formula C16H13BO2
Molecular Weight (g/mol) 248.088
MDL Number MFCD09260454
InChI Key ICQAKBYFBIWELX-UHFFFAOYSA-N
Synonym 4-naphthalen-2-yl phenylboronic acid, 4-2-naphthyl phenylboronic acid, 4-naphthalen-2-yl phenyl boronic acid, 4-2-naphthyl benzeneboronic acid, 4-naphthalen-2-yl phenylboronicacid, 4-2-naphthyl phenyl boronic acid, 4-naphthalen-1-yl-boronic acid, 4-naphth-1-yl benzeneboronic acid
PubChem CID 45382258
IUPAC Name (4-naphthalen-2-ylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)C2=CC3=CC=CC=C3C=C2)(O)O

Specifications

Quantity 200 mg
Formula Weight 248.09
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Naphthyl)phenylboronic Acid (contains varying amounts of Anhydride)