missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Morpholinoethoxy)aniline 98.0+%, TCI America™

Catalog No. M26915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26915G 5 g
M26911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26915G Supplier TCI America Supplier No. M26915G

Chemical Identifiers

CAS 52481-41-1
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.288
MDL Number MFCD04970981
InChI Key ZHFFNLQQANCJEQ-UHFFFAOYSA-N
Synonym 4-2-morpholinoethoxy aniline, 4-2-morpholin-4-ylethoxy aniline, 4-2-morpholin-4-yl-ethoxy-phenylamine, 4-2-morpholin-4-yl ethoxy aniline, 4-2-4-morpholinyl ethoxy aniline, 4-2-morpholin-4-yl-ethoxy phenylamine, 4-2-morpholin-4-ylethoxy phenylamine, 4-2-4-amino-phenoxy ethyl-morpholine
PubChem CID 6484711
IUPAC Name 4-(2-morpholin-4-ylethoxy)aniline
SMILES C1COCCN1CCOC2=CC=C(C=C2)N

Specifications

Melting Point 71°C
Color Yellow
Boiling Point 150°C
Quantity 5 g
Formula Weight 222.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Morpholinoethoxy)aniline