missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-4-thiazolyl)phenol 98.0+%, TCI America™

Catalog No. M23251G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23251G 1 g
M2325100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23251G Supplier TCI America Supplier No. M23251G
Only null left

Chemical Identifiers

CAS 30686-73-8
Molecular Formula C10H9NOS
Molecular Weight (g/mol) 191.248
MDL Number MFCD02642878
InChI Key HOAYTTLLVORNLA-UHFFFAOYSA-N
Synonym 4-2-methyl-4-thiazolyl phenol, 4-2-methyl-1,3-thiazol-4-yl phenol, 4-2-methylthiazol-4-yl phenol, 4-4-hydroxyphenyl-2-methylthiazole, 4-2-methyl-4-thiazolyl phenol for biochemical research, 4-2-methyl-thiazol-4-yl-phenol, phenol, p-2-methyl-4-thiazolyl, 4-2-methyl-4-thiazolyl phenol*, 4-2-methyl-1,3-thiazol-4-yl phenol #, phenol, 4-2-methyl-4-thiazolyl-, hydrochloride
PubChem CID 591743
IUPAC Name 4-(2-methyl-1,3-thiazol-4-yl)phenol
SMILES CC1=NC(=CS1)C2=CC=C(C=C2)O

Specifications

Melting Point 213°C
Color White-Yellow
Quantity 1 g
Formula Weight 191.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Methyl-4-thiazolyl)phenol [for Biochemical Research]