missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Methyl-4-thiazolyl)phenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 213°C |
| Color | White-Yellow |
| Quantity | 100 mg |
| Formula Weight | 191.25 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-(2-Methyl-4-thiazolyl)phenol [for Biochemical Research] |
Safety and Handling
| TSCA | No |