missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-1H-imidazol-1-yl)aniline, 97%, Thermo Scientific™

Numéro de catalogue. CC61414DE
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC61414DE 5g Amber Glass Bottle
CC61414DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC61414DE Fournisseur Thermo Scientific Chemicals Code fournisseur CC61414DE

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 74852-81-6
Formule moléculaire C10H11N3
Poids moléculaire (g/mol) 173.219
Numéro MDL MFCD06797788
Clé InChI IEZCMVRWKNEHJB-UHFFFAOYSA-N
Synonyme 4-2-methylimidazol-1-yl phenylamine, 4-2-methyl-1h-imidazol-1-yl aniline, 4-2-methyl-imidazol-1-yl-phenylamine, 4-2-methyl-1h-imidazol-1-yl benzenamine, 4-2-methylimidazol-1-yl aniline, 4-2-methyl-1h-imidazol-1-yl phenylamine, benzenamine, 4-2-methyl-1h-imidazol-1-yl, 4-2-methylimidazolyl phenylamine, pubchem19934, 1-4-aminophenyl-2-methylimidazole
PubChem CID 3157478
Nom de l’IUPAC 4-(2-methylimidazol-1-yl)aniline
SOURIRES CC1=NC=CN1C2=CC=C(C=C2)N

Spécifications

Nom ou substance chimique 4-(2-Methyl-1H-imidazol-1-yl)aniline
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 5g
Poids de formule 173.22
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.