missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)morpholine, 99%

Catalog No. p-7026649
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2275922 100 g
AAB2275930 250 g
AAB2275936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2275922 Supplier Thermo Scientific Chemicals Supplier No. B2275922

CAS: 622-40-2 | C6H13NO2 | 131.175 g/mol

4-(2-Hydroxyethyl)morpholine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Hydroxyethyl)morpholine is used as an organic chemical synthesis intermediate.

Solubility
Fully miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.

Chemical Identifiers

CAS 622-40-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00006180
InChI Key KKFDCBRMNNSAAW-UHFFFAOYSA-N
Synonym 2-morpholinoethanol, 4-2-hydroxyethyl morpholine, 4-morpholineethanol, n-2-hydroxyethyl morpholine, 2-4-morpholinyl ethanol, morpholine ethanol, morpholineethanol, hydroxyethylmorpholine, ethanol, 2-morpholino, beta-morpholinoethanol
PubChem CID 61163
ChEBI CHEBI:67144
IUPAC Name 2-morpholin-4-ylethanol
SMILES C1COCCN1CCO

Specifications

Melting Point 1°C to 2°C
Density 1.08
pH 10.7
Boiling Point 226°C to 228°C
Flash Point 99°C (210°F)
Refractive Index 1.4775
Quantity 100 g
Beilstein 104375
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material 4-(2-Hydroxyethyl)morpholine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-734-5
RTECSNumber QE3510000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only