missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Furyl)but-3-en-2-un, 97%, Thermo Scientific™

Numéro de catalogue. CD11326EA
Modifier l'affichage
Click to view available options
Quantité:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CD11326EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CD11326EA Fournisseur Thermo Scientific Chemicals Code fournisseur CD11326EA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 623-15-4
Formule moléculaire C8H8O2
Poids moléculaire (g/mol) 136.15
Numéro MDL MFCD00039566
Clé InChI GBKGJMYPQZODMI-SNAWJCMRSA-N
Synonyme furfurylideneacetone, 4-2-furyl-3-buten-2-one, 3-buten-2-one, 4-2-furanyl, fam polymer, oramin r, furfural acetone, oramin special gr, furfuralacetone polymer, polyfurfurylidineacetone, furfuralacetone
PubChem CID 735940
Nom de l’IUPAC (E)-4-(furan-2-yl)but-3-en-2-un
SOURIRES CC(=O)C=CC1=CC=CO1

Spécifications

Nom ou substance chimique 4-(2-Furyl)but-3-en-2-one
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 10 g
Poids de formule 136.15
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.