missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloro-6-nitrophenoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Numéro de catalogue. BTB02778EB
Modifier l'affichage
Click to view available options
Quantité:
1g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
BTB02778EB 25g
BTB02778DA 1g
BTB02778EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. BTB02778EB Fournisseur Thermo Scientific Chemicals Code fournisseur BTB02778EB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 175135-00-9
Formule moléculaire C12H7Cl2NO5S
Poids moléculaire (g/mol) 348.15
Numéro MDL MFCD00052679
Clé InChI CRTUVOFOPIFTQS-UHFFFAOYSA-N
Synonyme 4-2-chloro-6-nitrophenoxy benzene-1-sulfonyl chloride, 4-2-chloro-6-nitrophenoxy benzenesulfonyl chloride, 4-6-nitro-2-chlorophenoxy-benzenesulfonyl chloride, 4-2-chloro-6-nitrophenoxy benzene-1-sulphonyl chloride
PubChem CID 2774276
Nom de l’IUPAC 4-(2-chloro-6-nitrophenoxy)benzenesulfonyl chloride
SOURIRES C1=CC(=C(C(=C1)Cl)OC2=CC=C(C=C2)S(=O)(=O)Cl)[N+](=O)[O-]

Spécifications

Nom ou substance chimique 4-(2-Chloro-6-nitrophenoxy)benzene-1-sulfonyl chloride
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Quantité 25g
Poids de formule 348.16
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.