missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloro-6-nitrophenoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. p-2700549
Change view
Click to view available options
Quantity:
1g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB02778DA 1g
BTB02778EA 10g
BTB02778EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB02778DA Supplier Thermo Scientific Chemicals Supplier No. BTB02778DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 175135-00-9
Molecular Formula C12H7Cl2NO5S
Molecular Weight (g/mol) 348.15
MDL Number MFCD00052679
InChI Key CRTUVOFOPIFTQS-UHFFFAOYSA-N
Synonym 4-2-chloro-6-nitrophenoxy benzene-1-sulfonyl chloride, 4-2-chloro-6-nitrophenoxy benzenesulfonyl chloride, 4-6-nitro-2-chlorophenoxy-benzenesulfonyl chloride, 4-2-chloro-6-nitrophenoxy benzene-1-sulphonyl chloride
PubChem CID 2774276
IUPAC Name 4-(2-chloro-6-nitrophenoxy)benzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)Cl)OC2=CC=C(C=C2)S(=O)(=O)Cl)[N+](=O)[O-]

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 348.16
Percent Purity 97%
Chemical Name or Material 4-(2-Chloro-6-nitrophenoxy)benzene-1-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.