missing translation for 'onlineSavingsMsg'
Learn More

4-(2-bromoéthyle)tétrahydropyran, 97%, Thermo Scientific™

Numéro de catalogue. CC67408DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC67408DA 1 g Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC67408DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC67408DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 4677-20-7
Formule moléculaire C7H13BrO
Poids moléculaire (g/mol) 193.08
Numéro MDL MFCD09800414
Clé InChI WPDAWFCZGSQOPZ-UHFFFAOYSA-N
Synonyme 4-2-bromoethyl tetrahydropyran, 4-2-bromoethyl tetrahydro-2h-pyran, 4-2-bromoethyl oxane, 4-2-bromoethyl-tetrahydropyran, 4-2-bromo-ethyl-tetrahydro-pyran, 2h-pyran, 4-2-bromoethyl tetrahydro, 4-2-bromoethyl-tetrahydro-2h-pyran, pubchem16818, ablock ab-11-0011, 2h-pyran,4-2-bromoethyl tetrahydro
PubChem CID 22637012
Nom de l’IUPAC 4-(2-bromoéthyl)oxane
SOURIRES BrCCC1CCOCC1

Spécifications

Nom ou substance chimique 4-(2-Bromoethyl)tetrahydropyran
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 193.08
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.