missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Benzoxazolyl)-4'-(5-methyl-2-benzoxazolyl)stilbene 95.0+%, TCI America™

Catalog No. B156525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B156525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B156525G Supplier TCI America Supplier No. B156525G

Chemical Identifiers

CAS 5242-49-9
Molecular Formula C29H20N2O2
Molecular Weight (g/mol) 428.491
InChI Key SOTPOQQKAUOHRO-BQYQJAHWSA-N
PubChem CID 5761588
IUPAC Name 2-[4-[(E)-2-[4-(1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-5-methyl-1,3-benzoxazole
SMILES CC1=CC2=C(C=C1)OC(=N2)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)C5=NC6=CC=CC=C6O5

Specifications

Color Yellow
Quantity 25 g
Formula Weight 428.49
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Benzoxazolyl)-4′-(5-methyl-2-benzoxazolyl)stilbene
EINECSNumber (5)-6068
TSCA Yes