missing translation for 'onlineSavingsMsg'
Learn More

4-(2,6-Dichloro-4-pyrimidyl)morpholine 98.0+%, TCI America™

Catalog No. p-7128344
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45711G 1 g
D45715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45711G Supplier TCI America Supplier No. D45711G

Chemical Identifiers

CAS 52127-83-0
Molecular Formula C8H9Cl2N3O
Molecular Weight (g/mol) 234.08
MDL Number MFCD09746259
InChI Key QGGYMWHOBGSQCF-UHFFFAOYSA-N
Synonym 2,4-Dichloro-6-(4-morpholino)pyrimidine
PubChem CID 11586699
IUPAC Name 4-(2,6-dichloropyrimidin-4-yl)morpholine
SMILES C1COCCN1C2=CC(=NC(=N2)Cl)Cl

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 1 g
Formula Weight 234.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2,6-Dichloro-4-pyrimidyl)morpholine