missing translation for 'onlineSavingsMsg'
Learn More

4-([2,2' :6',2-Terpyridine″]-4'-yl)acide benzoïque 98,0+%, TCI America™

Numéro de catalogue. p-7137738
Modifier l'affichage
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T3405200MG 200 mg
T34051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T3405200MG Fournisseur TCI America Code fournisseur T3405200MG

Chemical Identifiers

CAS 158014-74-5
Molecular Formula C22H15N3O2
Molecular Weight (g/mol) 353.381
MDL Number MFCD22056799
InChI Key RPRRPKJKACRJDD-UHFFFAOYSA-N
Synonym 4′C-(4-Carboxyphenyl)-2,2′C:6′C,2′C′C-terpyridine
PubChem CID 15903886
IUPAC Name 4-(2,6-dipyridine-2-ylpyridine-4-yl)acide benzoïque
SMILES C1=CC=NC(=C1)C2=CC(=CC(=N2)C3=CC=CC=N3)C4=CC=C(C=C4)C(=O)O

Specifications

Melting Point 338°C
Color Blanc-jaune
Quantity 200 mg
Formula Weight 353.38
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 4-([2,2′:6′,2′′-Terpyridin]-4′-yl)benzoic Acid