missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrrol-1-yl)benzonitrile, 97%, Thermo Scientific™

Numéro de catalogue. CC25516EB
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC25516EB 25g
CC25516DA 1g
CC25516DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC25516EB Fournisseur Thermo Scientific Chemicals Code fournisseur CC25516EB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 23351-07-7
Formule moléculaire C11H8N2
Poids moléculaire (g/mol) 168.20
Numéro MDL MFCD00085164
Clé InChI OKVSZRKKRHNDOL-UHFFFAOYSA-N
Synonyme 4-1h-pyrrol-1-yl benzonitrile, 1-4-cyanophenyl pyrrole, n-4-cyanophenyl pyrrole, benzonitrile, 4-pyrrol-1-yl, 4-1-pyrrolyl benzonitrile, 4-pyrrol-1-yl benzonitrile, 4-pyrrolylbenzenecarbonitrile, acmc-1cekp, maybridge1_001756, 1-4-cyanophenyl-pyrrole
PubChem CID 272424
Nom de l’IUPAC 4-pyrrol-1-ylbenzonitrile
SOURIRES N#CC1=CC=C(C=C1)N1C=CC=C1

Spécifications

Nom ou substance chimique 4-(1H-pyrrol-1-yl)benzonitrile
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Quantité 25g
Poids de formule 168.2
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.