missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-ylméthyl)benzylamine hydrochlorure , Tech., Thermo Scientific™

Numéro de catalogue. CC56063DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC56063DA 1 g Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC56063DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC56063DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 904696-62-4
Formule moléculaire C11H14ClN3
Poids moléculaire (g/mol) 223.70
Numéro MDL MFCD07690519
Clé InChI QUVMWCVEYSYIOW-UHFFFAOYSA-N
Synonyme 4-1h-pyrazol-1-yl methyl phenyl methanamine hydrochloride, 4-1h-pyrazol-1-ylmethyl benzylamine hydrochloride, 4-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 1-4-1h-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 1-4-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 4-pyrazolylmethyl phenyl methylamine, chloride, 4-1h-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 4-1h-pyrazol-1-yl methyl phenyl methanaminehydrochloride, 1-4-1h-pyrazol-1-yl methyl phenyl methanamine-hydrogen chloride 1/1
PubChem CID 16269819
Nom de l’IUPAC [4-(pyrazol-1-ylméthyl)phényl]méthanamine; Chlorhydrate
SOURIRES Cl.NCC1=CC=C(CN2C=CC=N2)C=C1

Spécifications

Nom ou substance chimique 4-(1H-pyrazol-1-ylmethyl)benzylamine hydrochloride
CAS % minimum 80.0
CAS % maximal 100.0
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 223.71
Pourcentage de pureté Tech.
Niveau Caractéristiques
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.