missing translation for 'onlineSavingsMsg'
Learn More

[4-(1H-Pyrazol-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Numéro de catalogue. CC18409DA
Modifier l'affichage
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
CC18409DA Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC18409DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC18409DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Spécifications

Nom ou substance chimique [4-(1H-Pyrazol-1-yl)phenyl]methanol
CAS 143426-49-7
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Formule moléculaire C10H10N2O
Quantité 1g
Numéro MDL MFCD02682057
Synonyme 4-1h-pyrazol-1-yl phenyl methanol, 4-pyrazol-1-yl-phenyl methanol, 4-pyrazol-1-ylphenyl methanol, 4-1-pyrazolyl phenyl methanol, 4-1-pyrazolyl benzylalcohol, 4-pyrazol-1-yl phenyl methanol, 4-1h-pyrazol-1-yl benzyl alcohol, benzenemethanol, 4-1h-pyrazol-1-yl, 4-pyrazolylphenyl methan-1-ol
Clé InChI MMGMLIHSHFKRDK-UHFFFAOYSA-N
SOURIRES OCC1=CC=C(C=C1)N1C=CC=N1
Nom de l’IUPAC (4-pyrazol-1-ylphenyl)methanol
Poids moléculaire (g/mol) 174.20
PubChem CID 2776480
Poids de formule 174.2
Pourcentage de pureté 97%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.