missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Imidazol-1-ylmethyl)-N-methylbenzylamine 1.5 oxalate 0.5 hydrate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 910036-84-9
Molecular Formula C14H17N3O4
Molecular Weight (g/mol) 291.307
MDL Number MFCD09064952
InChI Key AIQAEMDKVTYASZ-UHFFFAOYSA-N
Synonym oxalic acid; 4-imidazol-1-ylmethyl phenyl methyl methyl amine, oxalic acid-1-4-1h-imidazol-1-yl methyl phenyl-n-methylmethanamine 1/1
PubChem CID 24229491
IUPAC Name 1-[4-(imidazol-1-ylmethyl)phenyl]-N-methylmethanamine;oxalic acid
SMILES CNCC1=CC=C(C=C1)CN2C=CN=C2.C(=O)(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC43396CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC43396DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

910036-84-9
291.307
AIQAEMDKVTYASZ-UHFFFAOYSA-N
24229491
CNCC1=CC=C(C=C1)CN2C=CN=C2.C(=O)(C(=O)O)O
C14H17N3O4
MFCD09064952
oxalic acid; 4-imidazol-1-ylmethyl phenyl methyl methyl amine, oxalic acid-1-4-1h-imidazol-1-yl methyl phenyl-n-methylmethanamine 1/1
1-[4-(imidazol-1-ylmethyl)phenyl]-N-methylmethanamine;oxalic acid

Specifications

910036-84-9
100.0
C14H17N3O4
oxalic acid; 4-imidazol-1-ylmethyl phenyl methyl methyl amine, oxalic acid-1-4-1h-imidazol-1-yl methyl phenyl-n-methylmethanamine 1/1
CNCC1=CC=C(C=C1)CN2C=CN=C2.C(=O)(C(=O)O)O
291.307
345.34
4-(1H-imidazol-1-ylmethyl)-N-methylbenzylamine 1.5 oxalate 0.5 hydrate
94.5
95%
MFCD09064952
AIQAEMDKVTYASZ-UHFFFAOYSA-N
1-[4-(imidazol-1-ylmethyl)phenyl]-N-methylmethanamine;oxalic acid
24229491
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents