missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Imidazol-1-yl)benzenecarbothioamide, 97%, Thermo Scientific™

Numéro de catalogue. CC18518DA
Modifier l'affichage
Click to view available options
Quantité:
1g
10g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC18518DA 1g Amber Glass Bottle
CC18518EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC18518DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC18518DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 423769-74-8
Formule moléculaire C10H9N3S
Poids moléculaire (g/mol) 203.263
Numéro MDL MFCD03659704
Clé InChI QUJXJNDFQWCYLF-UHFFFAOYSA-N
Synonyme 4-1h-imidazol-1-yl benzenecarbothioamide, benzenecarbothioamide,4-1h-imidazol-1-yl, 4-imidazol-1-yl benzenecarbothioamide, amino 4-imidazolylphenyl methane-1-thione, 4-1h-imidazol-1-yl benzothioamide
PubChem CID 2776487
Nom de l’IUPAC 4-imidazol-1-ylbenzenecarbothioamide
SOURIRES C1=CC(=CC=C1C(=S)N)N2C=CN=C2

Spécifications

Nom ou substance chimique 4-(1H-imidazol-1-yl)benzenecarbothioamide
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 203.27
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.