missing translation for 'onlineSavingsMsg'
Learn More

4,16-Dibromo[2.2]paracyclophane, 98%

Catalog No. AAB2574503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2574503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2574503 Supplier Thermo Scientific Chemicals Supplier No. B2574503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96392-77-7 | C16H14Br2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96392-77-7
Molecular Formula C16H14Br2
MDL Number MFCD09953451

Specifications

Melting Point 249°C to 252°C
Quantity 1 g
Formula Weight 366.1
Percent Purity 98%
Chemical Name or Material 4,16-Dibromo[2.2]paracyclophane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only