missing translation for 'onlineSavingsMsg'
Learn More

4-[(1-Pyrrolidinyl)carbonyl]phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. P21051G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P21051G Supplier TCI America Supplier No. P21051G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-[(1-Pyrrolidinyl)carbonyl]phenylboronic Acid (contains varying amounts of Anhydride)
CAS 389621-81-2
Color White-Yellow
Molecular Formula C11H14BNO3
Quantity 1 g
MDL Number MFCD03411950
Synonym 4-pyrrolidine-1-carbonyl phenylboronic acid, 4-pyrrolidine-1-carbonyl phenyl boronic acid, 4-pyrrolidinylcarbonylphenylboronic acid, 4-pyrrolidine-1-carbonyl benzeneboronic acid, 4-boronophenyl pyrrolidin-1-yl methanone, 1-4-boronobenzoyl pyrrolidine, boronic acid, 4-1-pyrrolidinylcarbonyl phenyl, 4-borono-n-1-pyrrolidinyl benzamide, 4-pyrrolidin-1-ylcarbonyl phenyl boronic acid
InChI Key VKPBESPVHGDDJS-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(C=C1)C(=O)N1CCCC1
IUPAC Name [4-(pyrrolidine-1-carbonyl)phenyl]boronic acid
Molecular Weight (g/mol) 219.05
PubChem CID 2773571
Formula Weight 219.05
Physical Form Crystalline Powder
Show More Show Less