missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)benzeneboronic acid pinacol ester, 97%

Catalog No. AAH59911MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59911MD 250 mg
AAH5991103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59911MD Supplier Thermo Scientific Chemicals Supplier No. H59911MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 852227-90-8 | C16H24BNO2 | 273.18 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 852227-90-8
Molecular Formula C16H24BNO2
Molecular Weight (g/mol) 273.18
MDL Number MFCD08060504
InChI Key DWJNNJSONWFVBT-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid pinacol ester, 4-1-pyrrolidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenylboronic acid pinacol ester, 4-pyrrolidin-1-yl benzeneboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl pyrrolidine
PubChem CID 18525715
IUPAC Name 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCC1

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 273.18
Percent Purity 97%
Chemical Name or Material 4-(1-Pyrrolidinyl)benzeneboronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only