missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)-1-butylamine, 98%

Catalog No. AAH5032306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5032306 5 g
AAH5032303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5032306 Supplier Thermo Scientific Chemicals Supplier No. H5032306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24715-90-0 | C8H18N2 | 142.25 g/mol

4-(1-Pyrrolidinyl)-1-butylamine is used directly as drugs, food, cosmetics. Used as a building block in organic synthesis, pyrrolidine activates ketones and aldehydes toward nucleophilic addition by formation of enamines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(1-Pyrrolidinyl)-1-butylamine is used directly as drugs, food, cosmetics. Used as a building block in organic synthesis, pyrrolidine activates ketones and aldehydes toward nucleophilic addition by formation of enamines

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 24715-90-0
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.25
MDL Number MFCD00192591
InChI Key LSDYCEIPEBJKPT-UHFFFAOYSA-N
Synonym 4-pyrrolidin-1-yl butan-1-amine, 4-1-pyrrolidinyl-1-butanamine, 1-pyrrolidinebutanamine, 4-1-pyrrolidino butylamine, 4-pyrrolidinobutylamine, 4-1-pyrrolidinyl-1-butylamine, 1-4-aminobutyl pyrrolidine, 4-pyrrolidin-1-yl-butylamine, 4-pyrrolidinylbutylamine, 1-pyrrolidinebutylamine
PubChem CID 2762762
IUPAC Name 4-pyrrolidin-1-ylbutan-1-amine
SMILES NCCCCN1CCCC1

Specifications

Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Soluble in water
Formula Weight 142.25
Percent Purity 98%
Chemical Name or Material 4-(1-Pyrrolidinyl)-1-butylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only