missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Phenyl-1H-benzimidazol-2-yl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. P21581G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P21581G Supplier TCI America Supplier No. P21581G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-(1-Phenyl-1H-benzimidazol-2-yl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 952514-79-3
Molecular Formula C19H15BN2O2
Quantity 1 g
MDL Number MFCD11045057
Synonym 4-(1-Phenyl-1H-benzimidazol-2-yl)benzeneboronic Acid
InChI Key PBSIVXAPTBHFFV-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C2=NC3=CC=CC=C3N2C4=CC=CC=C4)(O)O
IUPAC Name [4-(1-phenylbenzimidazol-2-yl)phenyl]boronic acid
Molecular Weight (g/mol) 314.151
PubChem CID 53249779
Formula Weight 314.15
Physical Form Crystalline Powder
Show More Show Less