missing translation for 'onlineSavingsMsg'
Learn More

[4-(1,3-Thiazol-2-yl)phenyl]methanol, 97%, Thermo Scientific™

Catalog No. cc40209da
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC40209DA 1g Amber Glass Bottle
CC40209DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC40209DA Supplier Thermo Scientific Chemicals Supplier No. CC40209DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 454678-91-2
Molecular Formula C10H9NOS
Molecular Weight (g/mol) 191.248
MDL Number MFCD09064950
InChI Key QOLUYHARONOGNO-UHFFFAOYSA-N
Synonym 4-1,3-thiazol-2-yl phenyl methanol, benzenemethanol,4-2-thiazolyl, 4-thiazol-2-yl benzyl alcohol, 4-thiazol-2-yl phenyl methanol, 4-1,3-thiazol-2-yl benzyl alcohol, 4-thiazol-2-yl benzylalcohol, 4-1,3-thiazol-2-yl phenyl methan-1-ol
PubChem CID 24229487
IUPAC Name [4-(1,3-thiazol-2-yl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)C2=NC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 191.25
Percent Purity 97%
Chemical Name or Material [4-(1, 3-Thiazol-2-yl)phenyl]methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.