missing translation for 'onlineSavingsMsg'
Learn More

[4-(1,3-Thiazol-2-yl)phenyl]methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC40209DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC40209DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

454678-91-2
191.248
QOLUYHARONOGNO-UHFFFAOYSA-N
24229487
C1=CC(=CC=C1CO)C2=NC=CS2
C10H9NOS
MFCD09064950
4-1,3-thiazol-2-yl phenyl methanol, benzenemethanol,4-2-thiazolyl, 4-thiazol-2-yl benzyl alcohol, 4-thiazol-2-yl phenyl methanol, 4-1,3-thiazol-2-yl benzyl alcohol, 4-thiazol-2-yl benzylalcohol, 4-1,3-thiazol-2-yl phenyl methan-1-ol
[4-(1,3-thiazol-2-yl)phenyl]methanol

Specifications

[4-(1, 3-Thiazol-2-yl)phenyl]methanol
96.5
97%
C10H9NOS
MFCD09064950
QOLUYHARONOGNO-UHFFFAOYSA-N
[4-(1,3-thiazol-2-yl)phenyl]methanol
24229487
97%
454678-91-2
100.0
Amber Glass Bottle
5g
4-1,3-thiazol-2-yl phenyl methanol, benzenemethanol,4-2-thiazolyl, 4-thiazol-2-yl benzyl alcohol, 4-thiazol-2-yl phenyl methanol, 4-1,3-thiazol-2-yl benzyl alcohol, 4-thiazol-2-yl benzylalcohol, 4-1,3-thiazol-2-yl phenyl methan-1-ol
C1=CC(=CC=C1CO)C2=NC=CS2
191.248
191.25
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents