missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ 3dGRO™ Human iPSC Derived Colon Organoids

Catalog No. SCC300MI
Change view
Click to view available options
No. of Cells:
≥ 200 Organoids/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Cells
SCC300MI ≥ 200 Organoids/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC300MI Supplier MilliporeSigma™ Supplier No. SCC300

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Highly characterized cryopreserved human colon intestinal organoids derived from iPSCs

  • Derived from integration-free human iPS cells reprogrammed using the Simplicon RNA reprogramming technology
  • Human iPS cells were differentiated using a multi-step process from definitive endoderm to posterior hind-gut endoderm and eventually into colon organoids

Specifications

Content And Storage Store in liquid nitrogen
Cell Type Organoid
For Use With (Application) Cell Culture, Stem Cell Culture
No. of Cells ≥ 200 Organoids/Vial

For research purposes only