3000ml Reaction Vessel, Cylindrical, 150mm Schott O-Ring Flange, 160mm O.D. x 280mm Height. Cylindrical reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. For clamp see CG-141 or CG-141-T, for o-ring see CG-147.
Numéro de catalogue.
CG192004
Fournisseur
Chemglass IncCode fournisseur
CG192004
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
3000ml Reaction Vessel, Cylindrical, 150mm Schott O-Ring Flange, 160mm O.D. x 280mm Height. Cylindrical reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. For clamp see CG-141 or CG-141-T, for o-ring see CG-147.