missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethoxysilyl)propyl Methacrylate (stabilized with BHT) 98.0+%, TCI America™

Catalog No. p-7132322
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M072525ML 25 mL
M0725100ML 100 mL
M0725500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M072525ML Supplier TCI America Supplier No. M072525ML

Chemical Identifiers

CAS 2530-85-0
Molecular Formula C10H20O5Si
Molecular Weight (g/mol) 248.35
MDL Number MFCD00008593
InChI Key XDLMVUHYZWKMMD-UHFFFAOYSA-N
Synonym 3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester
PubChem CID 17318
IUPAC Name 3-trimethoxysilylpropyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC[Si](OC)(OC)OC

Specifications

Color Colorless
Boiling Point 110°C
Quantity 25 mL
Formula Weight 248.35
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trimethoxysilyl)propyl Methacrylate (stabilized with BHT)
EINECSNumber (2)-2076
RTECSNumber UC0230000
TSCA Yes